WebSeal Well Plates by Thermo Scientific
A comprehensive range of PP well plates.
Our plate portfolio contains well plates which meets the requirements of today’s chromatographer using autosampler for plates. Compatible with a broad range of instruments, our well plate range offers solutions from simple routine analyses to specific and challenging sample handling problems.
Handle More Samples in Less Time
- Low volume and high recovery cavity
Solutions for all Applications
- Chromatography tested for solvent compatibility and inertness
Autosampler Compatible
- Using ANSI/SBS standards
Product Selection
Standard Plates and Mats
Low cost, standard performance commodity plates, seals and tapes (GC and LC tested and confirmed to be acceptable).
Certified Plates and Mats
Certified products, low bleed plates & high performance mats (GC/MS and LC/MS tested and confirmed to be excellent).
High Performance Plates and Mats
“Like working with a glass vial” high performance product range (surface modified plates; plates with real glass inserts, highest quality, products for high end chromatography; meet MSCERT quality requirements).