Bottles and Accessories
Made with the highest quality resins, Nalgene™ bottles and closures are engineered to work together with a proprietary valve seal and a strong, semi-buttress thread design. Our bottles offer a leakproof guarantee because we manufacture and test both components together as part of our routine quality program.
Product Selection
Bottles and Accessories
Our leakproof Nalgene bottles set the industry standard for quality and durability. Available in many sizes, styles and resins, we have a bottle to provide peace-of-mind service in your application.
Dropper Bottles
Our leakproof Nalgene bottles set the industry standard for quality and durability. Available in many sizes, styles and resins, we have a bottle to provide peace-of-mind service in your application.
Jars
Durable plastic jars are ideal for collection, transport or storage of solids, powders or pastes. Wide mouth allows easy access. Shatterproof.
Wash Bottles
Ensure safe liquid handling on your lab bench with Nalgene squeezable plastic wash bottles. Specially-labeled and color-coded versions are available for safe containment of chemicals and other hazardous substances