Spectrum offers BASF catalysts
BASF built on its world-renowned background in materials science and surface chemistry to create a range of efficient, high-performance precious metal powder catalysts. Its patented NanoSelect™ catalysts are defined by unimodal, nano-sized metal particles, increasing the available surface area of the metals to greatly boost catalytic activity. The innovative technology combines stabilizing and reducing functions to produce colloids that can be used with different support materials to create heterogeneous catalysts with unique catalytic behavior.
Product Selection
Platinum catalysts
Pre-reduced platinum, deposited to enable exceptionally favorable metal availability.
Palladium catalysts
Pre-reduced palladium, deposited to enable exceptionally favorable metal availability.
Rhodium catalysts
Pre-reduced rhodium, deposited to enable exceptionally favorable metal availability.
Ruthenium catalysts
Pre-reduced ruthenium, deposited to enable exceptionally favorable metal availability.