PIG® Hazmat Absorbents - ACID SPILLS? THINK PINK.
PIG® Hazmat Mats, rolls, socks & pillows are specially treated to soak up spills without degrading or reacting to corrosive liquids including:
- Sulfuric acid
- Hydrochloric acid
- Nitric acid
- Hydrofluoric acid
- Phosphoric acid
- Sodium hydroxide
- Potassium hydroxide
- Hydrogen peroxide
STRONGEST MAT ON THE MARKET
Eight layers of 100% polypropylene are thermally bonded to make PIG Mat the strongest mat on the market. It won’t rip, tear or fray, even when saturated.
OUTPERFORMS THE COMPETITION
Only PIG HazMat Absorbents are built to handle the worst of the worst. Highly corrosive liquids like 98% sulfuric acid and 30% sodium hydroxide can cause inferior cellulose mats, socks and pillows to degrade or react. But PIG is specially treated to absorb high concentrations of chemicals.
THE CLEAR CHOICE
Pink color is easily distinguishable to ensure workers use the correct mat during a spill emergency.
PIG® HazMat Chemical Absorbent Mat Pad
Lightweight 15" x 20", 200 pads. Safely soak up acids, bases, corrosives and hazardous chemical spills with our HazMat absorbent pads.
PIG® HazMat Chemical Absorbent Mat Pad
Heavyweight 15" x 20", 100 pads. Safely soak up acids, bases, corrosives and hazardous chemical spills with our HazMat absorbent pads.
PIG® HazMat Chemical Absorbent Mat Pad
Heavyweight 15"W x 20"L, 100 pads, 11 gal./bag. Safely soak up acids, bases, corrosives and hazardous chemical spills with our HazMat absorbent pads.
PIG® HazMat Chemical Absorbent Mat Pad in Dispenser Box
Heavyweight, 15" x 20", 100 pads. Safely soak up acids, bases, corrosives and hazardous chemical spills; dispenser box is easy to store.
PIG® HazMat Chemical Absorbent Mat Pad in Dispenser Box
Heavyweight, 10" x 13", 100 pads. Safely soak up acids, bases, corrosives and hazardous chemical spills; dispenser box is easy to store.
PIG® HazMat Chemical Absorbent Mat Roll
Heavyweight, 30" x 150', 1 roll. Safely soak up acids, bases, corrosives and hazardous chemical spills with our HazMat absorbent mat roll.
PIG® HazMat Chemical Absorbent Mat Roll
Heavyweight, 15" x 150', 1 roll. Safely soak up acids, bases, corrosives and hazardous chemical spills with our HazMat absorbent mat roll.
PIG® Rip-&-Fit® HazMat Chemical Absorbent Mat Roll
Heavyweight, 15" x 150', 1 roll. Create custom mat sizes to absorb chemical drips and spills. Just "rip" it along the perfs and "fit" it into place.
PIG® HazMat Chemical Absorbent Dike
5" x 10', Absorbs up to 9 gal. per sock, 2 socks. Contain and absorb higher-volume spills of concentrated chemicals.
PIG® HazMat Chemical Absorbent Sock
3" x 10', Absorbs up to 2 gal. per sock, 6 socks. Contain and absorb small spills of concentrated chemicals.
PIG® HazMat Chemical Absorbent Sock
3" x 46", Absorbs up to 100 oz. per sock, 12 socks. Contain and absorb small spills of concentrated chemicals.
PIG® HazMat Chemical Absorbent Pillow in Dispenser Box
Absorbs up to 30 oz. per pillow, 20 pillows. Our high-capacity HazMat pillows deliver the absorbency you need.Absorbs Acids; Bases; Unknowns.
PIG® HazMat Chemical Absorbent Pillow
Absorbs up to 1 gal. per pillow, 10 pillows. Our high-capacity HazMat pillows deliver the absorbency you need.Absorbs Acids; Bases; Unknowns.
PIG® HazMat Chemical Absorbent Pillow
Absorbs up to 76 oz. per pillow, 10 pillows. Our high-capacity HazMat pillows deliver the absorbency you need.Absorbs Acids; Bases; Unknowns.
MRO HazMat Pads
Heavyweight 15"x19", 100 pds. Value priced MRO Hazmat roll - When the job doesn't require the quality of PIG Mat, use MRO to get the job done!
MRO HazMat Roll
Heavyweight 15"x150', 1 roll. Value priced MRO Hazmat roll - When the job doesn't require the quality of PIG Mat, use MRO to get the job done!