See how Pall products can put these criminals behind bars
Barry Backpressure beaten backPall Acrodisc® One syringe filters have the high effective filtration area that Acrodisc products are known for, making for less backpressure than other products. That makes filtration quick and easy, especially useful for preparation of multiple samples. |
|||
Alice and Patty stopped in their tracksCleaning-up and degassing mobile phases with Pall SolVac® extends column life while maintaining results quality and consistency. Compared to glass funnel filters, SolVac, with its patented magnetic seal, is easy to use, eliminates washing and transfer steps, and minimizes spillage potential by drawing directly from the solvent bottle. |
|||
No way through for Patty ParticulateWith its wwPTFE media, Acrodisc One is the next generation of universal filtration, and has been shown to extend column life by over 52x with no significant increases in backpressure, saving on column replacement. |
|||
Eddie Extractable eradicatedThe Acrodisc MS range is tailor-made for mass spectrometery applications. Its water-wettable PTFE membrane and polyethylene hardware ensure extremely low extractable levels, reducing ion suppression / enhancement. |