Density and Refractometers
METTLER TOLEDO’s wide range of digital benchtop and portable density meters measure density, specific gravity (SG) and other related values, such as alcohol concentration, BRIX, API degrees, Baumé and Plato with high precision and a short measuring time.
The digital bench top refractometers offer more possibilities, and are the best choice for a wide range of applications from routine Brix measurements at 20°C to refractive index determinations of bitumen at 100°C.
Current Promotions
- Receive up to a $400 cash-back rebate with your next METTLER TOLEDO Balance, pH Meter, or Analytical Instrument purchase.
- Trade in your old balance and receive a 15% cash back rebate with your new Balance purchase.
Product Selection
Excellence Density Meters
The all-rounder benchtop density meter for a variety of applications.
Excellence Benchtop Refractometers
Affordable density meters (specific gravity meters) for daily applications.
Handheld Density Meter
Take your density measurements on site or in the field.
Handheld Refractometer
Handheld meters for refractive index
Density Accessories
Valuable options to make your operation effortless, secure and fast.
RE Accessories
Accessories to improve and secure your workflow.