Masterflex® Food and Beverage Pump & Tubing Solutions
Proper pump and tubing acquisition choice is necessary for sanitary food production.
Masterflex’s pumps and tubing are capable of handling a wide variety of fluids, from hot water and vitamin mixtures to the thickest molasses.
Our pump heads are designed for easy tubing exchange, and most many pump drives feature washdown capabilities. Plus, all pumps and tubing are compliant with food and beverage dispensing Choose from a wide variety of tubing that meets industry requirements from the following regulatory bodies:
- USP Class VI
- FDA
- NSF
- EP
- 3A
Masterflex food and beverage pump uses
Masterflex peristaltic pumps and tubing are designed for:
- Dispensing pizza sauce
- Pump-dispensing salad dressing
- Washing glass
- Pumping ice cream
- Automatically cleaning drains
- Dispensing popcorn butter
- Vitamin A & D injections
Masterflex food and beverage pumps advantages
What makes Masterflex the ideal choice for food and beverage applications?
- High purity. Fluid media only contacts tubing, which simplifies maintaining compliance. Tubing can be sterilized by ethylene oxide, gamma radiation, or autoclave usage.
- Wide range of tubing materials. Our tubing is chemically compatible with a variety of industrial solutions.
- Easy cleanup. Cleaning just means changing tubing. Plus IP66 washdown drives available
- Self-priming. Suction can lift up to 8.8 m (29 ft).
- Handles viscous materials. Works with Transfer and dispense heartier liquids such as yogurts, ice cream, and particulate-laden foods.
- Reversible. Operate in both directions without disconnecting the tubing.
- Modular design. You can mix and match a large variety of pump heads, tubing, and drives to fit your specific needs
- Custom engineered service. Design our pumps and tubing to fit into your dispensing or analytical equipment.
Featured Masterflex food and beverage pumps
Dispensing Pump
Versatile multichannel pumps for accurate low-flow fluid delivery – with an intuitive touch-screen interface
Sanitary Sampling Pump
Precise dispensing at variable speeds - NEMA 4X housings stand up to process and washdown applications.
Additive/Concentrate Pump
Program for “on demand” flavor concentrate. Many OEM application opportunities as well
Peristaltic Pump Tubing
Masterflex and Ismatec precision extruded and pump tested formulations engineered for the highest reliability, compliance, and traceability
Sanitary Fittings and Clamps
Ensure secure, sanitary connections in critical process applications. Fittings and clamps in sizes needed from pilot to production
Learn more about pumping viscous foods or beverages
Masterflex experts are here to help with all your viscous food and beverage pumping needs.
Contact us today to learn more.