Base Stands and Cabinets
Complete your installation of a Labconco fume hood and biosafety cabinet by using REDISHIP for fast delivery for cabinets and stands. The wide range of Protector cabinets include standard cabinets, cabinets that are specifically applicable to acid storage and solvent storage.- With the wide variety of base stands from Labconco, you have a selection of stands that are A.D.A. compliant, telescoping base stands, and our most popular electric and manual hydraulic lift base stands. Made in the USA. See how Labconco stands and cabinets can complete your laboratory needs
Protector Standard storage cabinets
Protector Standard storage cabinets support laboratory hoods, ventilation stations and balance enclosures. These durable steel cabinets have attractive glacier white exteriors which complement laboratory casework. Storage cabinets suitable for any lab.
Protector Acid Storage Cabinets
Protector Acid Storage Cabinets safely store and vent acids and other corrosive liquids. Versatile and support all Labconco benchtop fume hoods.
Labconco storage cabinets are designed to keep laboratories safe when storing acids and other corrosive liquids.
Protector solvent storage cabinets
Protector solvent storage cabinets – safely store and vent solvents and other flammable liquids. These cabinets are available with dual manual-closing, non-locking doors or automatic, self-closing doors. Labconco storage cabinets are designed to keep laboratories safe when storing acids and other corrosive liquids.