Performance level 2: High-throughput analysis, When robustness and reproducibility are key
Be compliant-ready for your high-throughput
samples in regulated environments
Choose Thermo Scientific™ SureSTART™ Performance level 2 products, to ensure high quality data with an uninterrupted workflow in high throughput applications using GC, HPLC/UHPLC, single or triple quadrupole MS systems. Use these Level 2 vials for routine QA/QC applications, where the analyte levels are achievable or close to the method detection level. These products have very low metal content and extractables and leachables and conform to high standards so that you can meet your regulatory requirements. They are Specification Certified to ensure their physical dimensions and autosampler compatibility.
If you need products that support routine analyses, our Performance Level 2 products are here for you. Select the products with the two waves.
Within Performance Level 2 you will find a selection of kits (including our range of EPA and Scintillation), vials, caps,inserts, septa and stoppers.
- All products within this performance level are suitable for GC, HPLC/UHPLC, and mass spectrometry (MS) applications
- Volume sizes ranging from 0.3 mL to 20 mL covering limited sample volumes to headspace analyses
Specification certified
- All Thermo Scientific™ SureSTART™ vials are dimensionally verified to ensure autosampler compatibility
- Inspected by automated optical and gauge inspection to ensure critical dimensions and tolerances
- Vials meet all requirements of US, EU, JPN Pharmacopeia
- Uniform and defect-free
Lot traceable
- 100% traceable through the manufacturing process
- Certificate of Conformance available by lot number upon request
Performance level 2 products include:
SureSTART™ Vials/ Closures
Glass: Amber/ Clear
Screw 2mL /9mm Screw AVCS™ Caps
Snap 2mL /11mm Snap and Crimp Caps (Magnetic Steel and Aluminum)
Crimp 2mL /11mm Crimp Caps (Magnetic Steel and Aluminum)
Microvials Screw 1.7mL- High Recovery / 9mm Screw AVCS™ Caps
Inserts
Headspace Glass: Amber/Clear
Crimp 10mL/ 20mm Crimp Caps (Magnetic Steel and Aluminum)
Crimp 20mL / 20mm Crimp Caps (Magnetic Steel and Aluminum)
20mm Septas (for 10+20mL vials)
20m Stoppers (for 10+20mL vials)
Screw 4mL /13mm Screw Cap
Screw 10mL / 18 Precision Screw Caps
Screw 20mL / 18 Precision Screw Caps
Vial/Cap. Septa Kits
Specification Certified 2 mL SureSTOP™ Screw Vial / AVCS™ Cap Kits
EPA Certified Screw Vial/ Cap Kits
Specification Certified 20 mL Screw Clear Glass Vial and 24 mm Closed Top Screw Cap Kits for Scintillation
Crimpers, Decrimpers and Decapping
Crimpers, Decrimpers and Decapping Pliers
Accessories for Programmable Electronic High-Power Crimp Stations
Electronic crimpers and decrimping
SureSTART™ Well Plates, Mats, Sealing Tapes, Handheld Applicators
Certified WebSeal™ Plates, Plastic, Non-Coated, Nonsterile
WebSeal™ Mats and Sealing Tapes, Non sterile