Equipment
Heathrow Scientific’s line of benchtop equipment is designed focusing on the demands of today’s lab professional to improve productivity and workflow processes. With proven 99.7% reliability and backed by a five-year warranty, our equipment delivers on versatility and value.
We offer a modern design and innovation with our wide selection of centrifuges, magnetic stirrers, shakers, vortex mixers, electronic pipettes, colony counters and more. We continue to grow our laboratory equipment line with multiple new product releases every year that include first-of-their-kind innovations like our latest new product, the MagFuge®.
MagFuge® Centrifuge and Magnetic Stirrer Combination
The MagFuge® is the industry’s first high-speed centrifuge and magnetic stirrer, combined into one unit. This design innovation enables a cost savings for labs by eliminating the need to purchase two pieces of equipment—a stirrer and centrifuge—because the MagFuge does it all. The high-speed centrifuge is easy to use and designed to deliver maximum speed (12,500 rpm/9,783 x g) that is ideal for most protocols requiring fast spins with the versatility to handle 0.2, 0.5, 1.5, 2.0 and 5.0 mL tube sizes.
The high-performance magnetic stirrer is designed using high-quality rare-earth magnets, creating a robust magnetic coupling that significantly reduces the chance of spin-outs and is perfect for work with sensitive samples and protocols up to 3 l requiring aggressive mixing for full dilution.
Gusto® Fast Mini Centrifuge
This compact centrifuge is ideal for protocols calling for fast spins with a higher RCF requirement. Ideal for PCR, microfilter cell separation, clinical and diagnostic applications and HPLC protocols. Made of durable ABS with UV-resistant coating. Brushless motor for quiet operation and low vibration. Quick-spin button for momentary spin requirements, press and hold to operate, release to end spin cycle. Tool-free rotor exchange. Safety features: sample imbalance, tilt auto shut-off and interlock lid prevents opening while rotor is turning.
Digital Orbital Shaker
Create your own matrix for vessels of all shapes and sizes with Hi-Lo platform and bungee cords. Instantly adjusts to weight load changes and unbalanced loads with no-stop electronic motor feedback, allowing for truly reproducible results. Durable ABS construction suitable for use in incubators (excluding C02) and cold rooms within the stated temperature range. Soft start and shut down feature reduces spilling from full vessels. Designed to handle heavy loads up to 10 lbs (4.5 kg). Overload protection shut-off prevents overloading.
Magnetic Induction Stirrer
Superior functional design with advanced magnetic coil technology drives the spinning of the stir bar, maintaining continuous, constant speed for reproducible results. Magnetic coupling is optimized by using alternating current (AC) as its driving force and is strong enough to stir viscous solutions. Motor-less operation generates minimal heat ideal for stirring temperature-sensitive samples and environments for long periods. With no moving parts, produces less vibration and quieter operation over traditional magnetic stirrers.