Furnace Cap Hood
Furnace cap hoods are designed to capture the lead fumes from your MAS3 fusion or cupellation furnaces. Designed for fire assay applications, our furnace cap hoods have a 10” flange for connecting to existing duct work and are required to be connected to an appropriately sized dust collector and a scrubber as required (items sold separately, contact us for more information).
Furnace cap hoods are made of steel and white powder coated for a long-lasting finish (additional colours available on request). Furnace cap hoods for other models of furnaces are also available upon request.