Go with the FLOW | Smart. Effective. Innovative.
Flow Cytometry Preparation
Scienceware® Flowmi™ Cell Strainers provides fast, efficient filtering of small volume samples (up to 1000µl) while providing reduced risk of cytometer clogs.
Key Features and Benefits:
- Preserves volume when filtering small sample volumes
- Removes cell aggregates and creates uniform single cell suspensions
- Decreases the potential for clogging of FLOW or FACS* instruments by effectively filtering cellular debris
- Sterile, 40µm or 70µm porosity
- Fits a wide variety of 1000µl Pipette tips
Recommended for use with samples having a maximum concentration of 2 million cells/ml (40µm) or 4 million cells/ml (70µm)