Cleanroom Gloves
Cleanroom glove liners from Wells Lamont are used everyday to protect people and their environment. These products are made in the USA for high quality and reliable availability.
Cut resistant glove liner commonly used in cleanroom construction
The M281 glove liner offers ANSI 4 cut resistance while being rated at Category III for particulates. Highly reusable with great dexterity.
Low lint highly reusuable nylon liner
The M555 liner is rated at Category II for particulates and is reusable up to 30 times.
Sterile cut resistant low lint glove
The M321 is a US Made and sterilized glove liner that offers ANSI A4 cut resistance and tests at Category III for particulates.