BRAND Titrette Bottletop
The BRAND Titrette provides safety and convenience to titration, eliminating meniscus reading error, while providing the precision of Class A glass burettes. Suitable for many common titrants, the Titrette is the first choice for routine titration in food and beverage labs, water/wastewater labs, petrochemical labs—anywhere where titration is part of a QC process. A recirculation valve helps purge bubbles, while included amber windows help protect light sensitive titrants. Fully disassebles for maintenance and cleaning. Three sizes - 10 mL, 25 mL and 50 mL, all available with optional RS232 interface to eliminate transcription error.
Product Selection
Titrette Bottletop Burette
Suitable for many common titrants up to 1 mol/liter in concentration