Reagent Reservoirs
Biotix disposable reagent reservoirs are designed for use with multi-channel pipetting applications and can accommodate up to 12-channel pipettes. Angled sides and a V-shaped bottom allow maximum recovery of reagents. All reservoirs feature internal graduated volume lines and pour-spouts on the corners. Choose from a wide variety of packaging options and styles.
- Most chemically resistant reservoir for use with a wide range of laboratory protocols
- Society for Biomolecular Sciences (SBS) footprint for use with automated instrumentation in addition to manual applications
- Precision engineered pour spouts and liquid collection channels for precise reagent handling
Available in three unique designs
Single well |
3-well divided |
Automation |