Solutions, VWR Chemicals BDH
The VWR Chemicals BDH line of solutions is designed with convenience in mind. The range includes common acid, base, and salt solutions in a variety of concentrations. Packaging sizes range from 500mL to 20L, in HDPE, glass, and Cubitainer® containers. Applications include:
- Titrations
- Environmental Analysis
- Quality Control
- ASTM/APHA Methods Testing