CE Pre-Prepared Trolley
Highest performance for the best contamination control!
The CE Pre-Prepared Trolley has been specifically designed and developed for use in controlled environments where maximum contamination control is needed. The trolley is electro-polished stainless steel with air tight connections and smooth welds. This ensures excellent biological control while being easy to clean and autoclave. The trolley is extremely functional and versatile with superior ergonomics and offers ample space for all utensils needed while cleaning effectively. Can be customized with various buckets, boxes, sieves, and bin bags.