Human Recombinant TPO (from E. coli)
Supplier: SARTORIUS CORPORATION EDI
CYK-0025-1012
CYK-0100-1012
77870-804EA
477.16
CAD
77870-804
77870-857
Human Recombinant TPO (from E. coli)
Proteins and Peptides
Manufactured to the highest quality standards using recombinant DNA technology which do not contain any animal-derived components or contaminants.