CentriVap® Cold Traps, –84 °C, 230 V
Supplier: Labconco
8611140 8611150 8611170 8611190 8611110 8611115 8611130
78007-998EA 15065.3 CAD
78007-998 78008-000 78008-002 78008-004 78007-992 78007-994 78007-996
CentriVap® Cold Traps, –84 °C, 230 V
Vacuum Traps
CentriVap Cold Traps are critical components designed to collect evaporated liquid from CentriVap Vacuum Concentrators. By condensing and trapping volatile solvents, they protect the vacuum pump and the laboratory environment from the harmful effects of vapors, fumes, and corrosives, while also ensuring that solvents can be easily collected for analysis or disposal.
  • 230 V models, not for use in USA
  • For low eutectic point samples containing water or acetonitrile
These traps are available either as integral components within configured CentriVap concentration systems or separately, making them suitable for a wide range of laboratory applications such as freeze drying and desiccation. Their role in maintaining system efficiency, safety, and solvent recovery makes them an essential part of many lab workflows.
Caution:
230 V models, not for use in USA. Corresponding models with voltage suitable for use in US (78007-990) are also available, please see separate product page.