CentriVap® Pro Acid-Resistant Vacuum Concentrators, 230 V
Supplier: Labconco
8512210 8512215 8512230 8512240 8512250 8512270 8512290 8512110 8512115 8512130 8512140 8512150 8512170 8512190
78007-860EA 21543.3 CAD
78007-860 78007-862 78007-864 78007-866 78007-868 78007-870 78007-872 78007-844 78007-846 78007-848 78007-850 78007-852 78007-854 78007-856
CentriVap® Pro Acid-Resistant Vacuum Concentrators, 230 V
Concentrators
The Acid-Resistant CentriVap® Pro vacuum concentrators features PTFE-coated aluminum chamber with Hastelloy shaft and other acid-resistant components with solvent resistant glass lid.
  • 230 V models, not for sale in USA
  • 5” full color touchscreen display
  • Rapidly concentrates multiple samples with corrosive chemicals
  • Durable acid-resistant components
  • Rotor imbalance detection
  • Unlimited user-set programs
  • High sample capacity
  • VaporWorks OS and Pre-programmed methods
  •  
Optional heat boost: Additional 300 W Heat Boost speeds chamber heating for faster sample processing and increased performance.
This device supports in-run and programmed parameter changes for full manipulation of rotor speed, chamber temperature from off to 100 °C, chamber preheat, run time and vacuum pressure. It features a pre-programmed menu of methods for a wide variety of sample types, making it versatile for different applications. The desiccant mode for vacuum desiccation of samples and Lyo-mode for small scale benchtop lyophilization when combined with a rotary vane vacuum pump and cold trap enhances its functionality for both drying and lyophilization processes.
To assist with sample processing, the system includes end point assist aids in determining dryness on select sample volumes and temperatures. Ethernet connectivity enables automated alerts about sample run data, method parameters, maintenance, and software updates, ensuring smooth operation and timely notifications. An automatic vacuum release valve and built-in vacuum delay allows rotor to achieve speed before applying vacuum contribute to operational efficiency.
User safety and convenience are prioritized with an audible alarm to signal completion of set point run time, a lid latch with safety sensor, and a phenol-free lid gasket. The centrifugal motion provided by a brushless motor featuring customizable rotor speed from 500 to 1800 rpm ensures precise control, while the 300 W chamber heater speeds evaporation for faster results.
The unit is built with a brushed stainless steel and charcoal, powder-coated steel exterior with blue accents. It includes a ¹/₂” OD polypropylene hose fitting for connection to vacuum tubing (vacuum pump not included), a vacuum port with quick disconnect fitting for ¹/₄” ID tubing, and an electrical receptacle on the back for connection to an accessory vacuum pump. It comes with a one year warranty on materials and workmanship.
Ordering information:
Full one year warranty.
Caution:
230 V models, not for use in USA. Corresponding models with voltage suitable for use in US (78007-858, 78007-842) are also available, please see separate product page.