Q5® Hot Start High-Fidelity DNA Polymerase
Supplier: New England Biolabs (NEB)
M0493L M0493S
CA102500-144EA 872 CAD
CA102500-144 CA102500-146
Q5® Hot Start High-Fidelity DNA Polymerase
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Q5 Hot Start High-Fidelity DNA Polymerase is a high-fidelity, thermostable, hot start DNA polymerase with 3' to 5' exonuclease activity, fused to a processivity-enhancing Sso7d domain to support robust DNA amplification.
  • Highest fidelity amplification (~280X higher than Taq)
  • Ultra-low error rates
  • Superior performance for a broad range of amplicons (from high AT to high GC)
  • Hot start and master mix formats available
The addition of an aptamer-based inhibitor allows room temperature reaction setup. With an error rate ~280-fold lower than that of Taq DNA Polymerase, Q5 Hot Start High-Fidelity DNA Polymerase is ideal for cloning and can be used for long or difficult amplicons. Q5 Hot Start High-Fidelity DNA Polymerase is supplied with an optimized buffer system that allows robust amplification regardless of GC content. The 5X Q5 Reaction Buffer contains 2 mM Mg++ at final (1X) reaction concentrations and is recommended for most routine applications. For GC-rich targets (≥65% GC), amplification can be improved by the addition of the 5X Q5 High GC Enhancer. Q5 Hot Start High-Fidelity DNA Polymerase is unlike typical, lower fidelity PCR enzymes.