BRAND® Transferpette® S -8/-12 Multi-Channel Mechanical Pipettes, Trade-Up Promotion

Supplier: Brandtech

705900TU 705912TU 705910TU 705908TU 705906TU 705920TU 705932TU 705930TU 705928TU 705926TU
MP76383-100EA 1034.19 CAD
MP76383-100 MP76383-102 MP76383-104 MP76383-106 MP76383-108 MP76383-110 MP76383-112 MP76383-114 MP76383-116 MP76383-118
BRAND® Transferpette® S -8/-12 Multi-Channel Mechanical Pipettes, Trade-Up Promotion
Pipettes Micropipettes

Transferpette® S pipettes are lightweight, ergonomic, fully autoclavable, and work with a wide range of tip brands.



  • True one-hand operation for right- and left-handed users; easily set the volume, pipette, and eject the tip

  • Color code on volume adjustment wheel, shaft, and volume-change protection

  • Low operating forces and a short stroke ensure relaxed pipetting, even during long pipetting series

  • Easy insertion and ejection of tips due to O-rings on multi-channel pipettes

  • Universal tip cone

  • Fully autoclavable at 121 °C


The volume-change protection is easy to operate with gloves on, and protects the set volume against accidental adjustment. The Transferpette® S also features Easy Calibration technology, which makes calibration a breeze. Calibration can be carried out without tools. A compatibility table shows which tips fit which device, so that the correct pipette tips can always be ordered.



Delivery information:

Supplied with performance certificate, battery, power adapter, reagent reservoir, installation tool, and silicone grease.


Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR