BRAND® Transferpette® -8/-12 Multi-Channel Electronic Pipettes, Trade-Up Promotion

Supplier: Brandtech

705420TU 705423TU 705426TU 705470TU 705473TU 705476TU 705419TU 705424TU 705469TU 705474TU 705398TU 705448TU
MP12000-506EA 1397.43 CAD
MP12000-506 MP12000-508 MP12000-510 MP12000-512 MP12000-514 MP12000-516 MP18900-518 MP18900-520 MP18900-522 MP18900-524 MP77322-324 MP77322-326
BRAND® Transferpette® -8/-12 Multi-Channel Electronic Pipettes, Trade-Up Promotion
Pipettes Micropipettes

Transferpette® electronic pipettes are lightweight, ergonomic, fully autoclavable, and work with a wide range of tip brands.



  • True one-hand operation for right- and left-handed users; easily set the volume, pipette, and eject the tip

  • Color code on volume adjustment wheel, shaft, and volume-change protection
  • Low operating forces and a short stroke ensure relaxed pipetting, even during long pipetting series
  • Easy insertion and ejection of tips due to O-rings on multi-channel pipettes
  • Universal tip cone
  • Fully autoclavable at 121 °C


The volume-change protection is easy to operate with gloves on, and protects the set volume against accidental adjustment. The Transferpette® electronic also features easy calibration technology, which makes calibration a breeze. Calibration can be carried out without tools. A compatibility table shows which tips fit which device, so that the correct pipette tips can always be ordered.



Delivery information:

Supplied with performance certificate, battery, power adapter, reagent reservoir, installation tool, and silicone grease.


Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR