Base Line Spill Pallets, Removable Grating, Forklift Access
Supplier: DENIOS INC
K16-1103 K16-3102 K16-3103 K16-1102
77609-826EA 1623.72 CAD
77609-826 77609-828 77609-830 77609-824
Base Line Spill Pallets, Removable Grating, Forklift Access
Drum Storage
Sumps are made from all-steel construction for an extended life. Feet for easy access to forklift in front and back.
  • Sumps are 100% tested and leak-free guaranteed  
  • Finished with a durable, hard-shell finish
  • For secondary drum containment
  • For 55 gallon drum containment
  • Constructed using premium 11-gauge quality steel
  • Choice of either galvanized or painted steel
Storage grate platforms are made from galvanized steel, they are removable for easy clean up in case of a spill. Great for oil, chemical, and universal secondary containment. This component provides superior performance in containment, ensuring full compliance with industry standards for spill prevention. It is designed for long-term durability and effective environmental protection, making it an ideal choice for a wide range of applications.
Certifications:
EPA, SPCC, OSHA, NFPA, UFC.