BrainStorm STEM Edu Energy Dragster Kit
Supplier: BRAINSTORM STUDIOS, LLC
SK200-0004 SK100-0004
470358-828PK 474 CAD
470358-828 470358-826
BrainStorm STEM Edu Energy Dragster Kit
Educational Materials Physics Educational Materials Mechanics Learning Activities
Build a rubber-band powered dragster to explore stored energy and motion.
  • Designed by teachers, for teachers
  • Demonstrates stored energy and propulsion
  • Perfect for home or the classroom
  • Available as a single kit or a class pack of ten with educator guide
  • No glue or mess
The BrainStorm energy dragster STEM kit engages students with the principles of kinetic and potential energy. Learners construct a lightweight dragster and power it using a wound rubber band to test speed and energy transfer. A fun, fast-paced way to investigate force and motion. These kits are authenticated as educational kits by STEM.org.
Delivery information:
Kit includes dragster parts, rubber bands, axle system, instruction guide.