Tornado Kit
Supplier: BRAINSTORM STUDIOS, LLC
SK100-0012 SK200-0012
470358-854EA 79 CAD
470358-854 470358-856
Tornado Kit
Educational Materials Earth Science Educational Materials Meteorology Learning Activities
Simulate a swirling vortex to explore fluid dynamics and weather systems.
  • Designed by teachers, for teachers
  • Demonstrates vortex formation and weather science
  • Perfect for home or the classroom
  • Available as a single kit or a class pack of ten with educator guide
  • No glue or mess
The BrainStorm Tornado STEM Kit allows students to simulate a mini tornado using a specially designed chamber. Learners explore the principles of fluid dynamics, rotation, and atmospheric pressure while creating controlled vortexes. A thrilling and educational way to introduce weather science. These kits are authenticated as educational kits by STEM.org.
Delivery information:
Kit includes vortex chamber, motor components, battery holder, instruction manual.