L-Glutamine Solution (200 mM), FUJIFILM Irvine Scientific
Supplier: FUJIFILM IRVINE SCIENTIFIC INC.
9317-100ML
9317-500ML
77975-381EA
60.44
CAD
77975-381
77975-382
L-Glutamine Solution (200 mM), FUJIFILM Irvine Scientific
L(+)-Glutamine
An essential amino acid important for cellular growth. Unstable at physiological pH.
- Animal-component free Chemically defined GMP manufacture
Formula:
C₅H₁₀N₂O₃ |
MDL Number:
MFCD00008044 CAS Number: 56-85-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...