Water for injection (WFI) Quality Water

Supplier: FUJIFILM IRVINE SCIENTIFIC INC.

9309-1L 9309-200L 9309-20L 9309-20LC
77975-376EA 54.45 CAD
77975-376 77975-377 77975-378 77975-379
Water for injection (WFI) Quality Water
Water

WFI Quality Water* meets or exceeds the testing requirements for sterile Water for Injection as stated in current USP, PhEur, and JP guidelines. Every WFI lot is tested according to USP, PhEur, and JP sterile Water for Injection standards and comes with a Certificate of Analysis.



WFI Quality Water is produced in accordance with the Guideline for Manufacture of In Vitro Diagnostic Products and Good Manufacturing Practices (GMPs) for medical devices. A variety of packaging sizes and volumes are available to support any use from research to high-throughput bioproduction.



Storage: Store at 2-30°C

Shelf Life: 24 months from date of manufacture


Formula: H₂O
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR