TubeWriter™ 400 UV Curable Inks
Supplier: TUBEWRITER, LLC
TW-INK-400 TW-UVB-1
77947-570EA 10163.35 CAD
77947-570 77947-572
TubeWriter™ 400 UV Curable Inks
Inks
TubeWriter's proprietary disposable UV curable ink is formulated to bond directly to any labware surface.
  • Bonds directly to any labware surface
  • Resistant to chemical solvents
  • Survives long-term storage at −80 °C and liquid nitrogen
  • Eliminates the need for sticky labels
  • Disposable cartridges with low replacement costs
To ensure the highest degree of solvent and temperatures protection possible, the ink has a shelf-life of 90 days.
The quarterly subscription delivers fresh ink cartridges every 90 days to ensure the highest degree of solvent and temperatures protection possible.