Static Rat/Hamster Cages, 2154F
Supplier: Tecniplast Usa
2154F025 2154F051 2F25B1000 ACBT0702 2154F001 2154F00SU 2154F114 2154F400SU ACBT0702SU 2154F014 1290D812 2F25B1000BW
75876-786EA 130.03 CAD
75876-786 75876-788 75876-794 75876-810 77322-422 77322-424 77322-426 77322-430 77969-766 77969-726 77969-690 77969-742
Static Rat/Hamster Cages, 2154F
Animal Supplies Animal Cages
The 2154F rat/hamster cage can comfortably house five rats of standard size and is available in polycarbonate or polysulfone materials.
  • Made from transparent plastic with exceptional impact strength and heat resistance
  • 146 square inch cage base
  • Compatible with a variety of lids, tops, accessories, and racks
  • Stainless steel accessories
  • Can house five rats of standard size
  • Racks come in a variety of sizes and can come with or without protective bumpers
The Tecniplast 2154F conventional cage is designed for housing laboratory rodents, particularly rats, offering a standard size and configuration that ensures compatibility with other equipment in animal research facilities. It features high-quality, durable materials for long-lasting use and easy maintenance.
The cage's smooth surfaces help reduce waste buildup, making cleaning more efficient while maintaining a hygienic environment for both animals and researchers.
With a focus on animal welfare, the 2154F cage provides a comfortable living space for the animals. Its design meets various animal welfare guidelines, offering enough space to reduce stress and enhance well-being.
The cage is also designed for ease of handling, simplifying maintenance and monitoring and making it a practical choice for research applications.
Accessories information:
All components and accessories are listed with the cage.
Standard lids are available with or without a hinged divider, which collapses to allow nesting of lids. Lids feature a built-in U-shaped feed hopper, solid sheet sides, swing-clip lock, and bottle protection washer, designed to prevent chewing of non-metal tops.
The removeable divider separates the built-in food hopper and the bottle holder. Most of the wire lids or raised wire lids, without hinged divider, can be equipped with removable dividers.
Metal bottle caps made of polished AISI 316 stainless steel are chew-proof and have a one-piece sipper tube design. The cone shaped cap design, with the slope of the bottle, maximizes water availability for animals.
Filter tops create a protective barrier at cage level with major advantages for the protection of both animals and staff. They can optionally be supplied with nylon lock latches to prevent accidental dislodgement from the cage body. Filter top cages can be autoclaved as complete units.
The raised bottom grid is made of AISI 304 stainless steel wire mesh and can be dropped into solid-bottom cages to separate animals from their waste. Grids feature 30 mm elevation for mice and for rats/guinea pigs and are easy to insert and remove.
Stainless steel racks are dedicated to husbandry and hygiene considerations, occupational health and safety requirements, ergonomics, reduced noise levels, and aesthetic appeal. Tubular base corners are all rounded for improved safety and to avoid damage of wall covering. Also available with four retrofitting plastic bumpers.
Ordering information:
Cage bases are offered in different materials. Order a cage base, lid, filter, top, bottle, bottle cap, divider, and raised bottom grid; all needed to build a full cage. To have enough material for cage change out processes, order 20 to 30% more than needed.
Delivery information:
Most of these items are stock items. Items that are out of stock will require additional time to ship.
Packaging:
Items are ordered individually and will be packaged to ensure safe and efficient transit.