Static Mouse Cage 1264C
Supplier: Tecniplast Usa
1264C025 1264C524 ACBT0402 ACBT0402SU 1264C016 1264C051 1264C400SUC
75876-744EA 78.97 CAD
75876-744 75876-746 75876-808 77969-762 77969-654 77969-656 77969-660
Static Mouse Cage 1264C
Animal Supplies Animal Cages
The 1264C mouse cage can comfortably house 3 mice of standard size and is available in polycarbonate or polysulfone materials.
  • Made from transparent plastic with exceptional impact strength and heat resistance
  • 57 square inch cage base
  • Compatible with a variety of lids, tops, accessories and racks
  • Stainless steel accessories
  • Can house 3 mice of standard size
The Tecniplast 1264C conventional cage is designed for housing laboratory rodents, particularly mice, offering a standard size and configuration that ensures compatibility with other equipment in animal research facilities. It features high-quality, durable materials for long-lasting use and easy maintenance. The cage's smooth surfaces help reduce waste buildup, making cleaning more efficient while maintaining a hygienic environment for both animals and researchers.
With a focus on animal welfare, the 1264C cage provides a comfortable living space for the animals. Its design meets various animal welfare guidelines, offering enough space to reduce stress and enhance well-being. The cage is also designed for ease of handling, simplifying maintenance and monitoring and making it a practical choice for research applications.
Accessories information:
All components and accessories are listed with the cage.
Standard lids are available with or without a hinged divider, which collapses to allow nesting of lids. Lids feature a built-in U-shaped feed hopper, solid sheet sides, swing-clip lock, and bottle protection washer, designed to prevent chewing of non-metal tops.
The removeable divider separates the built-in food hopper and the bottle holder. Most of the wire lids or raised wire lids, without hinged divider, can be equipped with removable dividers.
Metal bottle caps made of polished AISI 316 stainless steel are chew-proof and have a one-piece sipper tube design. The cone shaped cap design, with the slope of the bottle, maximizes water availability for animals.
Filter tops create a protective barrier at cage level with major advantages for the protection of both animals and staff. They can optionally be supplied with nylon lock latches to prevent accidental dislodgement from the cage body. Filter top cages can be autoclaved as complete units.
The raised bottom grid is made of AISI 304 stainless steel wire mesh, and can be dropped into solid-bottom cages to separate animals from their waste. Grids feature 30 mm elevation for mice and for rats/guinea pigs, and are easy to insert and remove.
Stainless steel racks are dedicated to husbandry and hygiene considerations, occupational health and safety requirements, ergonomics, reduced noise levels, and aesthetic appeal. Tubular base corners are all rounded for improved safety and to avoid damage of wall covering. Also available with four retrofitting plastic bumpers.
Ordering information:
Cage bases are offered in different materials. Order a cage base, lid, filter, top, bottle, bottle cap, divider, and raised bottom grid, all needed to build a full cage. To have enough material for cage change out processes, order 20 to 30% more than needed.
Delivery information:
Most of these items are stock items. Items that are out of stock will require additional time to ship.
Packaging:
Items are ordered individually and will be packaged to ensure safe and efficient transit.