HiTrap™ Protein Select™ Affinity Chromatography Columns
Supplier: Cytiva
17542151 17542152 17542153 17542154
CA77868-096EA 280.5 CAD
CA77868-096 CA77868-098 CA77868-100 CA77868-102
HiTrap™ Protein Select™ Affinity Chromatography Columns
Chromatography Columns
Cytiva™ Protein Select™ resin is available in HiTrap™ 1 and 5 ml columns, which are well suited for research, process development and scaling up to commercial scales.