Cole-Parmer® SHP-200 Undergrad Magnetic Hotplate Stirrer Kits
Supplier: Antyila Scientific
04807-93 04807-95 04807-97 04807-99
77971-906EA 1505.74 CAD
77971-906 77971-908 77971-910 77971-912
Cole-Parmer® SHP-200 Undergrad Magnetic Hotplate Stirrer Kits
Stirrers Magnetic Hotplate Stirrers
Offer accurate temperature control and overheating protection.
  • Compact, stackable design takes up less benchtop and storage space
  • Hot warning light remains on even when unplugged
  • Built with tough aluminum casing for longevity
  • Designed to prevent accidental spillage on control panel
  • BioCote® antimicrobial protection is built in for a cleaner, more hygienic product
These kits feature a magnetic hotplate stirrer designed with microprocessor-controlled dual thermocouples, providing accurate temperature control and protection from overheating. Powerful magnets and motor provide stirring speeds of up to 2000 rpm and mixing capability of up to 15 L of liquid. The square plate design allows flexibility of vessel sizes and combinations. To keep you safe, a 'hot' warning light flashes whenever the plate surface temperature is above 50 °C and continues to flash until the temperature drops below 50 °C, even when unplugged.
These stirring hotplates feature BioCote® antimicrobial protection which reduces the presence of odor and stain causing microbes such as bacteria, mold, and fungi on the product surface by up to 99.99%.  All the magnetic hotplate stirrers include an integral fitting for a retort rod and allow the retort base to slide underneath. For convenience and power saving, the units are built with a standard mains switch. To achieve accurate sample temperature control up to 200 °C (392 °F), the temperature controller can be connected to the unit and held in place with the probe holder and support rod, which are all included in the kit.
Ordering information:
Three year warranty.