Static Mouse Cages, 1144B
Supplier: Tecniplast Usa
ACCP2521 1144B001 1144B00SU 1144B116 1144B400SU ACBT0152 1144B016 1144B150 1144B400SUC 1264C812 2B64B1000 2B64B1000BW
95042-954EA 79.16 CAD
95042-954 77322-360 77322-364 77322-366 77322-368 77969-760 77969-634 77969-636 77969-638 77969-662 77969-734 77969-736
Static Mouse Cages, 1144B
Animal Supplies Animal Cages
The 1144B Mouse Cage can comfortably house 3 mice of standard size, and is available in polycarbonate or polysulfone materials.
  • Made from transparent plastic with exceptional impact strength and heat resistance
  • 52 sq.in. cage base
  • Compatible with a variety of lids, tops, accessories, and racks
  • Stainless steel accessories
  • Can house three mice of standard size
  • Racks come in a variety of sizes, with or without protective bumpers
The Tecniplast 1144B conventional cage is designed for housing laboratory rodents, particularly mice, offering a standard size and configuration that ensures compatibility with other equipment in animal research facilities. It features high-quality, durable materials for long-lasting use and easy maintenance. The cage's smooth surfaces help reduce waste buildup, making cleaning more efficient while maintaining a hygienic environment for both animals and researchers.
With a focus on animal welfare, the 1144B cage provides a comfortable living space for the animals. Its design meets various animal welfare guidelines, offering enough space to reduce stress and enhance well being. The cage is also designed for ease of handling, simplifying maintenance and monitoring, and making it a practical choice for research applications.
Accessories information:
All components and accessories are listed with the cage.
Standard lids are available with or without a hinged divider, which collapses to allow nesting of lids. Lids feature a built-in U-shaped feed hopper, solid sheet sides, swing-clip lock, and bottle protection washer, designed to prevent chewing of non-metal tops.
The removeable divider separates the built-in food hopper and the bottle holder. Most of the wire lids or raised wire lids, without hinged divider, can be equipped with removable dividers.
Metal bottle caps made of polished AISI 316 stainless steel are chew-proof and have a one-piece sipper tube design. The cone shaped cap design, with the slope of the bottle, maximizes water availability for animals.
Filter tops create a protective barrier at cage level with major advantages for the protection of both animals and staff. They can optionally be supplied with nylon lock latches to prevent accidental dislodgement from the cage body. Filter top cages can be autoclaved as complete units.
The raised bottom grid is made of AISI 304 stainless steel wire mesh, and can be dropped into solid-bottom cages to separate animals from their waste. Grids feature 30mm elevation for mice and for rats/guinea pigs, and are easy to insert and remove.
Stainless steel racks are dedicated to husbandry and hygiene considerations, occupational health and safety requirements, ergonomics, reduced noise levels, and aesthetic appeal. Tubular base corners are all rounded for improved safety and to avoid damage of wall covering. Also available with four retrofitting plastic bumpers.
Ordering information:
Cage bases are offered in different materials. Order a cage base, lid, filter, top, bottle, bottle cap, divider, and raised bottom grid; all needed to build a full cage. To have enough material for cage change out processes, order 20-30% more than needed.
Delivery information:
Most of these items are stock items. Items that are out of stock will require additional time to ship.
Packaging:
Items are ordered individually and will be packaged to ensure safe and efficient transit.