Ward's® Lichen Slides
Supplier: Avantor
470176-858EA 9.4 CAD
470176-858 470181-102 470181-104 470181-106
Ward's® Lichen Slides
Slides Prepared Slides
A composite of fungi and algae.
  • Ward's produced slides
  • Rigorous quality control standards
A great example of mutualism, lichen are a composite organism of fungi and algae. In these slides both parts of this relationship are easily seen.