Ward's® Plant Comparison Slides
Supplier: Avantor
470176-980EA 11.85 CAD
470176-980 470176-982 470177-478 470177-480 470177-482 470177-484 470177-486 470181-428 470181-432 470181-434 470181-436
Ward's® Plant Comparison Slides
Slides Prepared Slides
Multiple sections on a single slide.
  • Ward's produced slides
  • Rigorous quality control standards
Easily compare structures in plant material with these comparison slides. Each slide contains multiple specimens to make comparison of structures easy. Leaf sections are typically arranged vertically, while stem and root slides are generally horizontal.
Order Now
SPECIFICATIONS
Environmentally Preferable