Ward's® Buttercup (Ranunculus) Slides
Supplier: Avantor
470176-880EA 13.45 CAD
470176-880 470177-470 470181-380 470181-384
Ward's® Buttercup (Ranunculus) Slides
Slides Prepared Slides
A herbaceous dicot.
  • Ward's produced slides
  • Rigorous quality control standards
Compare the anatomy of a common dicot. Ranunculus is most often used to show root development. The large cortex and central vascular cylinder make tissue identification easy.