Ward's® Moss (Mnium) Slides
Supplier: Avantor
470176-862EA 45.75 CAD
470176-862 470176-864 470177-826 470177-828 470181-122 470181-124 470181-130
Ward's® Moss (Mnium) Slides
Slides Prepared Slides
Common moss with a mixed life cycle.
  • Ward's produced slides
  • Rigorous quality control standards
Mosses are a familiar organism for students, and an easily found bryophyte. These pioneer bryophytes grow in many different conditions, and transform areas with poor growing conditions. Mosses produce through an alternating life cycle. The most visible stage is the haploid vegetative form. Individual gametophytes form and fertilization occurs in an archegonia forming a diploid zygote. The zygote divides into many haploid spores as the archegonia forms an
More Product Information