Dry Block Heaters, Analog
Supplier: Ohaus
30392045 30392052 30392066 30392073
77874-088EA 698.44 CAD
77874-088 77874-090 77874-094 77874-096
Dry Block Heaters, Analog
Block Heaters Analogue Block Heaters
Versatile Dry Block Heaters are ideal for applications that require temperature stability.
  • Painted steel housing, support rod holder with locking knob
  • Dual-adjustment knobs with dial markings
  • All units hold one optional interchangeable modular block with over 40 choices
  • Blocks are easy to use, no tools required
Close tube-to-block contact enables maximum heat retention, resulting in efficient heating. Analog models are an economical choice.
Applications: Incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling
More Product Information