PVC Stand for Powder and Liquid Samplers
Supplier: GLOBEPHARMA
STD.10.PVC STD.22.PVC STD.42.PVC
77887-188EA 248.46 CAD
77887-188 77887-306 77888-042
PVC Stand for Powder and Liquid Samplers
Samplers Solid Samplers
Unit-dose and Bulk Powder, Liquid and Semi-solid Samplers available in various lengths and configurations.
  • Precision Sampling: Designed for accurate and repeatable sampling of powders from bulk containers, ensuring representative and consistent samples for quality control and testing
  • Pharmaceutical-Grade Construction: Made from high-quality 316 L stainless steel or FDA-compliant materials, ensuring compatibility with GMP environments and resistance to corrosion or contamination
  • Ease of Cleaning and Sterilization: Engineered for quick disassembly and
More Product Information