VWR® Acid/Corrosive Storage Cabinets, Blue
Supplier: VWR International
77530-092 VWRC145 VWRC160 VWRC330 VWRC345 VWRC360
77530-092EA 2499.64 CAD
77530-092 77530-094 77530-096 77530-098 77530-100 77530-102
VWR® Acid/Corrosive Storage Cabinets, Blue
The leak tight sump and shelves of the cabinet are equipped with removable corrosive-resistant trays to contain accidental spills.
  • 18-Gauge double wall construction and seamed joints
  • Double wall construction with 1½" air space
  • Leak proof sill, 2" deep
  • Non-sparking , three-point, self-latching doors
  • Static grounding connection
  • Large warning label
Acid cabinets are finished in corrosive-resistant blue urethane paint. Large warning label reads: "CAUTION CORROSIVES".
Certifications:
Complies with current OSHA regulations, FM-factory mutual approved, NFPA, designed in accordance with specifications set forth by the National Fire Protection Association.
Accessories information:
Extra shelves and trays, explosion proof exhaust blower and motors, keys, casters, and self-closing kits are available.
Caution:
Cabinet by itself is not recommended for the storage of nitric or sulfuric acids unless used in conjunction with VWRPE3045 polyethylene cabinet.