Powder and Liquid Samplers, II
Supplier: GLOBEPHARMA
M2.10.SD.SS M2.20.SD.SS M2.3.3.SS M2.30.SD.SS M2.40.SD.SS M2.6B.CE.SS M2.HB.SD.SS M2.SB.SD.SS
77887-832EA 1105.62 CAD
77887-832 77887-404 77887-480 77887-370 77888-418 77887-358 77887-364 77887-312
Powder and Liquid Samplers, II
Samplers Solid Samplers
Unit-dose and bulk powder, liquid, and semi-solid samplers available in various lengths and configurations.
  • Precision sampling: designed for accurate and repeatable sampling of powders from bulk containers, ensuring representative and consistent samples for quality control and testing
  • Pharmaceutical-grade construction: made from high-quality 316 L stainless steel or FDA-compliant materials, ensuring compatibility with GMP environments and resistance to corrosion or contamination
  • Ease of cleaning and sterilization: engineered for quick disassembly and thorough cleaning, supporting clean-in-place (CIP) and autoclave sterilization procedures
  • Customizable and scalable: can be tailored to specific sampling applications with adjustable volumes and configurations, ideal for R&D, pilot, and production-scale environments
  • Compliance ready: supports regulatory compliance with FDA, USP, and ISO standards, making them suitable for validated sampling protocols in pharmaceutical and cleanroom operations
Global Pharma specializes in high-quality sampling solutions for the pharmaceutical, biotech, and related industries. The product portfolio includes unit dose and bulk powder, liquid, and semi-solid samplers, designed to meet stringent regulatory requirements. These samplers are available in a variety of configurations to ensure precise and contamination-free sample collection.
With a focus on innovation and compliance, Global Pharma’s samplers cater to diverse manufacturing environments, providing single-use and reusable options for enhanced process efficiency and sterility assurance. Their products are widely used in quality control, R and D, and production settings, making them a trusted choice for pharmaceutical manufacturers worldwide.