Powder and Liquid Samplers, MTCM-II
Supplier: GLOBEPHARMA
MTCM.II.5.DS3 MTCM.II.9.32.P MTCM.II.AS MTCM.II.B.UH MTCM.II.C.E.B MTCM.II.DS3.UH MTCM.II.10.B MTCM.II.10.DS3 MTCM.II.3.16.P MTCM.II.3.8.P MTCM.II.3.8D.P MTCM.II.5.16.P MTCM.II.5.B
77887-044EA 14894.88 CAD
77887-044 77887-418 77887-712 77888-024 77887-278 77887-088 77888-114 77888-496 77888-120 77887-962 77887-040 77887-024 77888-088
Powder and Liquid Samplers, MTCM-II
Samplers Solid Samplers
Unit-dose and bulk powder, liquid and semi-solid samplers available in various lengths and configurations.
  • Precision sampling: Designed for accurate and repeatable sampling of powders from bulk containers, ensuring representative and consistent samples for quality control and testing
  • Pharmaceutical-grade construction: Made from high-quality 316 L stainless steel or FDA-compliant materials, ensuring compatibility with GMP environments and resistance to corrosion or contamination
  • Ease of cleaning and sterilization: Engineered for quick disassembly and thorough cleaning, supporting clean-in-place (CIP) and autoclave sterilization procedures
  • Customizable and scalable: Can be tailored to specific sampling applications with adjustable volumes and configurations, ideal for R&D, pilot, and production-scale environments
  • Compliance ready: Supports regulatory compliance with FDA, USP, and ISO standards, making them suitable for validated sampling protocols in pharmaceutical and cleanroom operations
Global Pharma specializes in high-quality sampling solutions for the pharmaceutical, biotech, and related industries. Their product portfolio includes unit dose and bulk powder, liquid, and semi-solid samplers, designed to meet stringent regulatory requirements. These samplers are available in a variety of configurations to ensure precise and contamination-free sample collection.
With a focus on innovation and compliance, Global Pharma’s samplers cater to diverse manufacturing environments, providing single-use and reusable options for enhanced process efficiency and sterility assurance. Their products are widely used in quality control, R&D, and production settings, making them a trusted choice for pharmaceutical manufacturers worldwide.