Powder and Liquid Samplers, Model IC
Supplier: GLOBEPHARMA
M1.C.0.1.SD.SS M1.C.0.2.SD.SS M1.C.0.3.SD.SS M1.C.0.7.SD.SS M1.C.0.8.SD.SS M1.C.0.9.SD.SS M1.C.1.0.SD.SS M1.C.1.1.SD.SS M1.C.1.2.SD.SS M1.C.3.5.SD.SS M1.C.3.6.SD.SS M1.C.3.7.SD.SS M1.C.3.8.SD.SS M1.C.3.9.SD.SS M1.C.4.0.SD.SS M1.C.6.SS M1.C.2.5.SD.SS M1.C.2.6.SD.SS M1.C.2.7.SD.SS M1.C.2.8.SD.SS M1.C.2.9.SD.SS M1.C.3.0.SD.SS M1.C.3.1.SD.SS M1.C.3.2.SD.SS M1.C.3.3.SD.SS M1.C.3.4.SD.SS M1.C.0.4.SD.SS M1.C.0.5.SD.SS M1.C.0.6.SD.SS M1.C.1.7.SD.SS M1.C.1.8.SD.SS M1.C.1.9.SD.SS M1.C.12.SS M1.C.18.SS M1.C.2.0.SD.SS M1.C.2.1.SD.SS M1.C.2.2.SD.SS M1.C.2.3.SD.SS M1.C.2.4.SD.SS M1.C.1.3.SD.SS M1.C.1.4.SD.SS M1.C.1.5.SD.SS M1.C.1.6.SD.SS
77888-076EA 323 CAD
77888-076 77888-368 77887-262 77887-796 77887-682 77887-766 77887-064 77887-864 77887-806 77888-420 77888-462 77887-218 77887-360 77887-956 77888-092 77888-036 77888-174 77887-500 77887-308 77887-576 77888-494 77888-354 77887-640 77887-302 77888-122 77887-540 77887-920 77887-830 77888-046 77887-866 77887-052 77887-138 77887-594 77888-038 77888-060 77888-068 77887-036 77887-646 77887-804 77888-480 77887-978 77887-180 77887-760
Powder and Liquid Samplers, Model IC
Samplers Solid Samplers
Unit-dose and bulk powder, liquid, and semi-solid samplers available in various lengths and configurations.
  • Precision sampling: Designed for accurate and repeatable sampling of powders from bulk containers, ensuring representative and consistent samples for quality control and testing
  • Pharmaceutical-grade construction: Made from high-quality 316 L stainless steel or FDA-compliant materials, ensuring compatibility with GMP environments and resistance to corrosion or contamination
  • Ease of cleaning and sterilization: Engineered for quick disassembly and thorough cleaning, supporting clean-in-place (CIP) and autoclave sterilization procedures
  • Customizable and scalable: Can be tailored to specific sampling applications with adjustable volumes and configurations, ideal for R&D, pilot, and production-scale environments
  • Compliance ready: Supports regulatory compliance with FDA, USP, and ISO standards, making them suitable for validated sampling protocols in pharmaceutical and cleanroom operations
Global Pharma specializes in high-quality sampling solutions for the pharmaceutical, biotech, and related industries. The product portfolio includes unit dose and bulk powder, liquid, and semi-solid samplers, designed to meet stringent regulatory requirements. These samplers are available in a variety of configurations to ensure precise and contamination-free sample collection.
With a focus on innovation and compliance, Global Pharma’s samplers cater to diverse manufacturing environments, providing single-use and reusable options for enhanced process efficiency and sterility assurance. Their products are widely used in quality control, R&D, and production settings, making them a trusted choice for pharmaceutical manufacturers worldwide.