VWR® Autoclavable Bags, Polypropylene, Clear
Supplier: NAN SING PLASTICS LIMITED CA
0038021031VW02 77778-324 77778-330 77778-336 77778-400 77778-412 77778-414 77778-360 77778-362 77778-364 77778-366 77778-368 77778-378 77778-390 77778-444 77778-338 77778-344 77778-348 77778-350 77778-424 77778-434
77778-320CS 247.21 CAD
77778-320 77778-324 77778-330 77778-336 77778-400 77778-412 77778-414 77778-360 77778-362 77778-364 77778-366 77778-368 77778-378 77778-390 77778-444 77778-338 77778-344 77778-348 77778-350 77778-424 77778-434
VWR® Autoclavable Bags, Polypropylene, Clear
Bags Sterilization Pouches
Sturdy, temperature-resistant bags are ideal for disposal of non-hazardous waste that must first be autoclaved. Bags can also be used for autoclaving other products.
  • Autoclavable up to 135 °C (275 °F) maximum
  • Available in multiple thicknesses: 37.5 µm/1.5 mil or 50 µm/2 mil
  • Available plain or as a printed bag
  • Highly resistant to tears and punctures
Bags are supplied complete with 20 cm twist tie wire closures that allow expansion of the bag and ventilation when cooling. Bags meet the requirements of the 165 g puncture inspection standard as described in ASTM D1709.
Certifications:
ISO 9001:2015, Sedex.