VWR® Autoclavable Biohazard Bags, Polypropylene, Orange
Supplier: VWR International
77778-380 77778-388 77778-352 77778-422 77778-436
77778-380CS 258.35 CAD
77778-380 77778-388 77778-352 77778-422 77778-436
VWR® Autoclavable Biohazard Bags, Polypropylene, Orange
Bags Sterilization Pouches
Sturdy, temperature-resistant bags are ideal for disposal of non-hazardous waste or hazardous waste that must first be autoclaved. Bags can also be used for autoclaving other products.
  • Autoclavable up to 135 ℃ (275 ℉) maximum
  • Printed bags feature the universal biohazard symbol
  • Temperature indicator text ‘contents have been autoclaved’ turns dark to indicate successful autoclaving
  • Highly resistant to tears and punctures
Bags are supplied complete with 20 cm twist tie wire closures that allow expansion of the bag and ventilation when cooling. Bags meet the requirements of the 165 g puncture inspection standard as described in ASTM D1709.
Certifications:
ISO 9001:2015, Sedex.