VWR® Autoclavable Biohazard Bags, Polypropylene , Clear
Supplier: VWR International
77778-322 77778-328 77778-334 77778-402 77778-408 77778-372 77778-342
77778-322CS 481.35 CAD
77778-322 77778-328 77778-334 77778-402 77778-408 77778-372 77778-342
VWR® Autoclavable Biohazard Bags, Polypropylene , Clear
Bags Sterilization Pouches
Sturdy, temperature-resistant bags are ideal for disposal of non-hazardous waste or hazardous waste that must first be autoclaved. Bags can also be used for autoclaving other products.
  • Autoclavable up to 135 ℃ (275 ℉) maximum
  • Available in multiple thicknesses: 37.5 µm/1.5 mil or 50 µm/2 mil
  • Printed bags feature the universal biohazard symbol
  • Temperature indicator text ‘contents have been autoclaved’ turns dark to indicate successful autoclaving
  • Highly resistant to tears and punctures
Bags are supplied complete with 20 cm twist tie wire closures that allow expansion of the bag and ventilation when cooling. Bags meet the requirements of the 165 g puncture inspection standard as described in ASTM D1709.
Certifications:
ISO 9001:2015, Sedex.