Cleanroom Tables, Heavy Duty
Supplier: PALBAM 2008 AC LTD TE
WX1208 WX1210
77898-330EA 4160.49 CAD
77898-330 77898-260
Cleanroom Tables, Heavy Duty
Furniture Tables and Workbenches
Palbam’s cleanroom heavy duty tables and table accessories are precision-engineered stainless steel workstations designed for demanding controlled environments.
  • Heavy-duty construction: Made from high-grade stainless steel (typically 304/316L), ensuring superior durability, corrosion resistance, and long-lasting performance in critical environments
  • Cleanroom-compliant design: Smooth, crevice-free surfaces with fully welded seams minimize particle accumulation, meeting ISO 14644 standards for cleanliness
  • Customizable configurations: Available with optional accessories such as undershelves, drawers, casters, and adjustable leveling feet to fit various applications and space constraints
  • Ergonomic and efficient: Designed with user comfort in mind, promoting better posture and productivity during long periods of use
  • Static dissipative options: ESD-safe models available for electronics and microelectronics environments to prevent static discharge and protect sensitive components
  • Enhanced workflow support: Integrated accessories and modular design streamline workstation setup, improving efficiency and organization in gowning or processing areas
These durable, ergonomic tables offer maximum stability and load-bearing capacity, making them ideal for tasks requiring a clean and robust workspace. Featuring a sleek, easy-to-clean design that meets stringent cleanroom standards, they are available in a variety of sizes and configurations.
Accessories information:
Accessories such as undershelves, drawers, and adjustable feet enhance functionality and adaptability. Palbam’s tables support optimized workflows and ensure compliance in critical cleanroom operations.
Ordering information:
For optional ESD-safe models (sold separately); contact your Avantor representative. 