VWR® LH-5000 Homogenizers

Supplier: VWR International
77898-152 77898-154
77898-152EA 3634 CAD
77898-152 77898-154
VWR® LH-5000 Homogenizers
Homogenizers Handheld Homogenizers

The VWR® LH-5000  is a powerful 850 W homogenizer that was specifically designed to disrupt a wide range of samples in volumes ranging from 200 µl to 5 L.



  • Powerful 850 W motor

  • Variable speed from 5000 to 25000 rpm
  • Volume range from 200 µl to 5 L
  • Quiet operation
  • Compatible with disposable plastic generator probes
  • Wide range of stainless steel generator probes available from 5 to 30 mm in diameter


Certifications:

CE marked.


Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR