VION Water, ASTM Type I

Supplier: ALDON CORP SE

VN-WW7001-60ML VN-WW7001-125ML VN-WW7001-1L VN-WW7001-250ML VN-WW7001-500ML
470358-610EA 10.98 CAD
470358-610 470358-598 470358-608 470358-604 470358-614
VION Water, ASTM Type I
Chemicals for Science Education

CAS Number: 7732-18-5
Formula Weight: 18.02
Formula: H2O

Meets the strictest standards defined by the American Society for Testing and Materials (ASTM).



  • Reverse osmosis treated and deionized (RODI) water.

  • Ideal for a wide range of ultra-sensitive laboratory uses.


Vion ultrapure, ASTM Type I, water meets the strictest standards defined by the American Society for Testing and Materials (ASTM). It is ideal for a wide range of ultra-sensitive laboratory uses including gas chromatography, HPLC, mass spectrometry, atomic absorption spectroscopy, and trace analysis.



Specifications
Resistivity ≥ 18 MΩ
Conductivity ≤ 0.056 μS/cm
Total Organic Carbon (TOC) ≤ 50 ppb
Silicate (as SiO2) ≤ 3 ppb
Chloride (Cl) ≤ 1 ppb
Sodium (Na) ≤ 1 ppb
Endotoxin ≤ 0.03 EU/mL
Heterotrophic BacteriaCount (HBC) ≤ 10 CFU/L


Formula: H₂O
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR